This product is now discontinued, View Related Products

Salvislab Mixcenter Stirring Hot Plates

Smooth stirring action to 1500 rpm
  • Ceramic-coated stainless steel top plate offers even heating and good chemical resistance
  • Hot surface warning stays lit until top plate falls below 50°C even when heating function in turned off
  • Use with an external temperature probe for greater precision (probe purchased separately)
SalvisLab MixCenter is ideal for thermally demanding tasks. Available in both analog and digital models with precise temperature and stirring control, these stirring hot plates are built for all applications in the lab. Digital model has an RS-232 port for external PC control.
$1,046.48 - $1,824.06CAD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
SalvisLab 31A06036 Mixcenter MC35 Analog Stirring Hot Plate, 115 VAC RK-04646-03 Mfr # 31A06036
COMPARE
115
$1,046.48CAD / Each
SalvisLab Mixcenter MC35 Analog Stirring Hot Plate, 220 VAC RK-04646-04 Mfr # 31A06035
COMPARE
220
$1,119.78CAD / Each
SalvisLab Mixcenter MC350 Digital Stirring Hot Plate, 115 VAC RK-04646-05 Mfr # 31A06360
COMPARE
115
$1,824.06CAD / Each
SalvisLab Mixcenter MC350 Digital Stirring Hot Plate, 220 VAC RK-04646-06 Mfr # 31A06350
COMPARE
220
$1,824.06CAD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat