Safety Spark Lighters
Sure, safe ignition for your bunsen burner
Use this well-constructed, safety spark lighter for all your burner ignition needs. Special spark hood protects the flint. Gas pocket forms beneath the hood, ensuring instant lighting.
Lighter incorporates new larger, renewable flint that produces up to 1500 ignitions..
$17.09 - $50.10CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Safety Spark Laboratory Lighter with 5 Flints RK-36136-50 Mfr # 3021
COMPARE
Safety Spark Bunsen Burner/Laboratory Lighter with 5 Flints
$17.09CAD / Each
Porta-Lab PSL-2012 Piezo Spark-O-Matic Electronic Spark Lighter RK-36310-60 Mfr # PSL-2012
COMPARE
Piezo Spark-O-Matic Electronic Spark Lighter
$50.10CAD / Each