This product is now discontinued, View Related Products

Rubbermaid Platform Trucks

Ideal for moving large or bulky loads
  • Plastic truck won't rust, dent, or warp
Plastic platform trucks are maintenance-free and virtually indestructible—won't rust, dent, or warp like metal or wood trucks. Sturdy handle secures your load and offers easy mobility.
$1,116.83 - $1,502.86CAD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Capacity (lbs) Length (in) Width (in)
Availability Pricing
Rubbermaid 4403 Platform Truck, 36" x 24", 500 lb capacity RK-47200-00 Mfr # 4403BLA
COMPARE
500 24 36
$1,116.83CAD / Each
Rubbermaid 4406 Platform Truck, 36" x 24", 1000 lb capacity RK-47200-01 Mfr # 4406
COMPARE
1000 24 36
$1,236.17CAD / Each
Rubbermaid 4436 Platform Truck, 48" x 24", 1000 lbs Cap RK-47200-10 Mfr # 4436
COMPARE
1000 24 48
$1,319.42CAD / Each
Rubbermaid 4466-86 Platform Truck, 60" x 30", 1000 lbs Cap RK-47200-19 Mfr # 4466-86
COMPARE
1000 60
$1,368.78CAD / Each
Rubbermaid 4471 Platform Truck, 30" x 60", 2000 lbs Cap RK-47200-20 Mfr # 4471
COMPARE
2000 30 60
$1,502.86CAD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat