Cole-Parmer® Essentials Round-Bottom Lab Sampling Scoops, HDPE
Round shape facilitates easy transfer of samples
  • Suitable for food-grade applications
  • High resistance to chemicals and aqueous solutions of salts, acids, and alkalis
  • Reusable; provides a more sustainable option to disposable scoops
Useful in laboratory and rugged process applications, these smooth, round bottom lab scoops allow easy and complete transfer of sample materials. Made of high-density polyethylene (HDPE), they withstand exposure to most chemicals without corroding. These sampling scoops are suitable to be used with food-grade products.
When used in place of disposable scoops, these reusable lab scoops help reduce waste in the laboratory, as well as save you money and reordering time. Indicated capacities are approximate and are not intended for measuring specific volumes.
$110.50 - $200.38CAD / Case of 5
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Handle Length (cm) Overall Length (cm)
Availability Pricing
COMPARE
250 8.89 15.24
$110.50CAD / Case of 5
COMPARE
500 9.53 19.69
$134.08CAD / Case of 5
COMPARE
1000 11.43 24.13
$187.12CAD / Case of 5
COMPARE
1500 11.43 28.58
$200.38CAD / Case of 5