This product is now discontinued, View Related Products
Reticles
Add a scale to your microscope inspections for increased accuracy and precision
Designed to increase the accuracy of your inspections, these clear glass discs are marked with precise scales or patterns. Reticles fit into microscope eyepieces at the intermediate image area of the specimen or object being examined. Both the scale or pattern of the reticle and the object image are superimposed and in focus.
$157.65 - $190.07CAD / Each
28 variations of this product are available.
See All Product Options Below
28 PRODUCT OPTIONS
Compareitem specifications
Eyepiece Diameter (mm) Description
Availability Pricing
Klarmann Rulings KR-203-19MM Reticle, Scaled from 0.300, 19-mm diameter RK-46800-00 Mfr # KR-203-19MM
COMPARE
19 Reticle, Scaled from 0.300, 19-mm diameter
$190.07CAD / Each
Klarmann Rulings KR-207-19MM Reticle, Scaled from 10 mm/100 Divisions, 19 mm dia RK-46800-01 Mfr # KR-207-19MM
COMPARE
19 Reticle, Scaled from 10 mm/100 Divisions, 19 mm dia
$184.17CAD / Each
Klarmann Rulings KR-301-19MM Microscope Eyepiece Reticle, crossline pattern, 19 mm dia RK-46800-02 Mfr # KR-301-19MM
COMPARE
19 Microscope Eyepiece Reticle, crossline pattern, 19 mm dia
$159.13CAD / Each
Klarmann Rulings KR-406-19MM Microscope Eyepiece Reticle, grid pattern, 19 mm dia RK-46800-03 Mfr # KR-406-19MM
COMPARE
19 Microscope Eyepiece Reticle, grid pattern, 19 mm dia
$187.12CAD / Each
Klarmann Rulings KR-501-19MM Reticle, 10 Concentric Circles with Crossline, 19 mm dia RK-46800-04 Mfr # KR-501-19MM
COMPARE
19 Reticle, 10 Concentric Circles with Crossline, 19 mm dia
$160.60CAD / Each
Klarmann Rulings KR-613-19MM Microscope Eyepiece Reticle, cross scale pattern, 19 mm dia RK-46800-05 Mfr # KR-613-19MM
COMPARE
19 Microscope Eyepiece Reticle, cross scale pattern, 19 mm dia
$176.81CAD / Each
Klarmann Rulings KR-848-19MM Reticle, Comparator Pattern, 360° total, 19 mm dia RK-46800-06 Mfr # KR-848-19MM
COMPARE
19 Reticle, Comparator Pattern, 360° total, 19 mm dia
$160.60CAD / Each
Klarmann Rulings KR-203-21MM Reticle, Scaled from 0.300, 21-mm diameter RK-46800-10 Mfr # KR-203-21MM
COMPARE
21 Reticle, Scaled from 0.300, 21-mm diameter
$175.33CAD / Each
Klarmann Rulings KR-207-21MM Reticle, Scaled from 10 mm/100 Divisions, 21 mm dia RK-46800-11 Mfr # KR-207-21MM
COMPARE
21 Reticle, Scaled from 10 mm/100 Divisions, 21 mm dia
$184.17CAD / Each
Klarmann Rulings KR-301-21MM Microscope Eyepiece Reticle, crossline pattern, 21 mm dia RK-46800-12 Mfr # KR-301-21MM
COMPARE
21 Microscope Eyepiece Reticle, crossline pattern, 21 mm dia
$176.81CAD / Each
Klarmann Rulings KR-406-21MM Microscope Eyepiece Reticle, grid pattern, 21 mm dia RK-46800-13 Mfr # KR-406-21MM
COMPARE
21 Microscope Eyepiece Reticle, grid pattern, 21 mm dia
$184.17CAD / Each
Klarmann Rulings KR-501-21MM Reticle, 10 Concentric Circles with Crossline, 21 mm dia RK-46800-14 Mfr # KR-501-21MM
COMPARE
21 Reticle, 10 Concentric Circles with Crossline, 21 mm dia
$185.65CAD / Each
Klarmann Rulings KR-613-21MM Microscope Eyepiece Reticle, cross scale pattern, 21 mm dia RK-46800-15 Mfr # KR-613-21MM
COMPARE
21 Microscope Eyepiece Reticle, cross scale pattern, 21 mm dia
$160.60CAD / Each
Klarmann Rulings KR-848-21MM Reticle, Comparator Pattern, 360° total, 21 mm dia RK-46800-16 Mfr # KR-848-21MM
COMPARE
21 Reticle, Comparator Pattern, 360° total, 21 mm dia
$162.07CAD / Each
Klarmann Rulings KR-203-22.8MM Reticle, Scaled from 0.300, 22.8-mm diameter RK-46800-20 Mfr # KR-203-22.8MM
COMPARE
22.8 Reticle, Scaled from 0.300, 22.8-mm diameter
$163.55CAD / Each
Klarmann Rulings KR-207-22.8MM Reticle, Scaled from 10 mm/100 Divisions, 22.8 mm dia RK-46800-21 Mfr # KR-207-22.8MM
COMPARE
22.8 Reticle, Scaled from 10 mm/100 Divisions, 22.8 mm dia
$181.23CAD / Each
Klarmann Rulings KR-301-22.8MM Microscope Eyepiece Reticle, crossline pattern, 22.8 mm dia RK-46800-22 Mfr # KR-301-22.8MM
COMPARE
22.8 Microscope Eyepiece Reticle, crossline pattern, 22.8 mm dia
$175.33CAD / Each
Klarmann Rulings KR-406-22.8MM Microscope Eyepiece Reticle, grid pattern, 22.8 mm dia RK-46800-23 Mfr # KR-406-22.8MM
COMPARE
22.8 Microscope Eyepiece Reticle, grid pattern, 22.8 mm dia
$187.12CAD / Each
Klarmann Rulings KR-501-22.8MM Reticle, 10 Concentric Circles with Crossline, 22.8 mm dia RK-46800-24 Mfr # KR-501-22.8MM
COMPARE
22.8 Reticle, 10 Concentric Circles with Crossline, 22.8 mm dia
$160.60CAD / Each
Klarmann Rulings KR-613-22.8MM Reticle, Cross Scale Pattern, 22.8 mm dia RK-46800-25 Mfr # KR-613-22.8MM
COMPARE
22.8 Reticle, Cross Scale Pattern, 22.8 mm dia
$159.13CAD / Each
Showing 1 - 20 of 28