This product is now discontinued, View Related Products
Resealable Open-Head Drums
No more heavy steel drums
  • Withstand rough handling—lightweight, rugged design
  • Easy access through 2" and 3⁄4" NPT bungs
  • Tough sides can support mixer
Open-head drums feature a removable lid sealed by a neoprene gasket; a polycoated steel ring with threaded tightening bolt securely fastens lid to drum. Constructed of durable LLDPE, these drums are suitable for a wide range of applications.
$209.52 - $346.10CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Material Volume (Gal) Wall thickness (in)
Availability Pricing
COMPARE
LLDPE 15 0.12
$209.52CAD / Each
COMPARE
LLDPE 30 0.187
$264.18CAD / Each
COMPARE
LLDPE 55 0.25
$346.10CAD / Each