This product is now discontinued, View Related Products

Replacement Probes for Extech Video Inspection Boroscopes

Compatible with Extech Video Borescope/Wireless Inspection Camera
Enhance the versitility of your inspection camera with these optional camera heads available with longer cables.
$176.79 - $397.80CAD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Extech BR-9CAM-2M Replacement 9mm Borescope Camera Head With 2m Cable RK-68517-53 Mfr # BR-9CAM-2M
COMPARE
Replacement 9mm Borescope Camera Head With 2m Cable
$210.68CAD / Each
Extech BR-9CAM-5M Replacement 9mm Borescope Camera Head With 5m Cable RK-68517-55 Mfr # BR-9CAM-5M
COMPARE
Replacement 9mm Borescope Camera Head With 5m Cable
$278.46CAD / Each
Extech BR-17CAM Replacement 17mm Borescope Camera Head With 1m Cable RK-68517-57 Mfr # BR-17CAM
COMPARE
Replacement 17mm Borescope Camera Head With 1m Cable
$176.79CAD / Each
Extech BR-17CAM-2M Replacement 17mm Borescope Camera Head With 2m Cable RK-68517-59 Mfr # BR-17CAM-2M
COMPARE
Replacement 17mm Borescope Camera Head With 2m Cable
$210.68CAD / Each
Extech BR-17CAM-5M Replacement 17mm Borescope Camera Head With 5m Cable RK-68517-61 Mfr # BR-17CAM-5M
COMPARE
Replacement 17mm Borescope Camera Head With 5m Cable
$397.80CAD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat