Replacement Grid sets for Scienceware Switch-Grid Tube Racks
Less expensive than purchasing multiple racks
  • Color coded grid inserts fit 10 to 30mm test tubes
Easily switch out grids to hold other tube sizes as needed with the multiple grids that are available. Constructed of chemical resistant polyoxymethylene plastic the grids are easily cleaned with mild detergent, ethanol or isopropyl alcohol. Autoclavable at 250°F (121°C).
$11.49CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Max Tube Size (mm) Tubes Held
Availability Pricing
Scienceware F18745-1300 Grid Set for Switch-Grid Test Tube Rack, Holds 10-13mm Tubes, Orange. Pack of 4. RK-06736-18 Mfr # F18745-1300
COMPARE
Orange 13 84
$11.49CAD / Each
Scienceware F18745-1600 Grid Set for Switch-Grid Test Tube Rack, Holds 13-16mm Tubes, Blue. Pack of 4. RK-06736-19 Mfr # F18745-1600
COMPARE
Blue 16 60
$11.49CAD / Each
Scienceware F18745-2000 Grid Set for Switch-Grid Test Tube Rack, Holds 16-20mm Tubes, Green. Pack of 4. RK-06736-22 Mfr # F18745-2000
COMPARE
Green 20 40
$11.49CAD / Each
Scienceware F18745-2500 Grid Set for Switch-Grid Test Tube Rack, Holds 20-25mm Tubes, Fuchsia. Pack of 4. RK-06736-23 Mfr # F18745-2500
COMPARE
Fuchsia 25 24
$11.49CAD / Each
Scienceware F18745-3000 Grid Set for Switch-Grid Test Tube Rack, Holds 25-30mm Tubes, Yellow. Pack of 4. RK-06736-24 Mfr # F18745-3000
COMPARE
Yellow 30 12
$11.49CAD / Each