This product is now discontinued, View Related Products
Releasable Design Cable Ties
Reusable-work on a bundle without needing a new tie
  • Simply depress tab to remove tie
  • Withstand temperatures to 185°F (85°C)
These ties are injection-molded of 6/6 nylon— won’t corrode or stretch. Durable locking mechanism holds securely even when used to bundle vibrating or flexing materials.
Ties continue to hold tension even in oily spray or wet environments. Ties withstand temperatures to 185°F (85°C). All ties remain flexible to –40°F (–40°C). Efficient range of ties may be higher or lower in your specific application. Melting temperature is 495°F (257°C).
$327.09CAD / Bag of 1000
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max Bundle Diameter (in) Length (in) Tensile Strength (lbs)
Availability Pricing
COMPARE
3 11 50
$327.09CAD / Bag of 1000