This product is now discontinued, View Related Products
Red Lion Miniature Length Sensor
Spring-loaded torsion arm for quick installation
  • Compact size
  • Quadrature output
  • Built-in spring tensioning
  • Vertical, horizontal, or upside-down mounting for quick installation
Designed for light to medium duty sensing applications, the Miniature Length Sensor, model ZMD, is compact in size and easy-to-use. It features a built-in spring-loaded torsion arm that provides a simple-to adjust torsion load, allowing the unit to be mounted in almost any orientation, including upside down.
Using a 6" or 200 mm wheel, the ZMD can be used on almost any surface, while operating at speeds up to 3000 feet per minute. The housing is a durable, conductive composite material that will eliminate static build up. Whether you need to measure speed, position, or distances, the Model ZMD is the ideal solution.
$553.99CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Actuator Type Max Operating Temperature (° F) Max Supply Voltage (VDC)
Availability Pricing
Red Lion ZMD0250B Miniature Length Sensor Quad Output 250 PPR RK-68493-52 Mfr # ZMD0250B
COMPARE
250 PPR 185 28
$553.99CAD / Each
Red Lion ZMD1000B Miniature Length Sensor Quad Output 1000 PPR RK-68493-54 Mfr # ZMD1000B
COMPARE
1000 PPR 185 28
$553.99CAD / Each