This product is now discontinued, View Related Products

Red Lion G3 series HMI Operator Panels

Benchmark of reliability and durability
  • Models come standard with one 10/100Base-T(X) Ethernet, two RS-232, and one RS-422/485 serial ports.
  • Communicates in over 300 different protocols.
  • Built-in data loggers for continuous data, event, and alarm collection and monitoring.
  • Data is compiled in CSV-formated files and can be stored, displayed, emailed, or automatically synchronized with a server.
  • A web server is built-in to allow remote control and monitoring.
  • Programming via Red Lion Crimson 3.0 software platform.
$1,975.82 - $6,014.38CAD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Red Lion G310S210 HMI 10.4" TFT 32K VGA 640x480 LCD, Outdoor RK-30018-52 Mfr # G310S210
COMPARE
HMI 10.4\" TFT 32K VGA 640x480 LCD, Outdoor
$6,014.38CAD / Each
Red Lion G308A210 HMI 8.4" TFT 32K VGA 640x480 LCD, Indoor RK-30018-53 Mfr # G308A210
COMPARE
HMI 8.4\" TFT 32K VGA 640x480 LCD, Indoor
$3,950.16CAD / Each
Red Lion G306A000 HMI 5.7" TFT Active Matrix QVGA 320x240 LCD, Indoor RK-68515-47 Mfr # G306A000
COMPARE
HMI 5.7\" TFT Active Matrix QVGA 320x240 LCD, Indoor
$1,975.82CAD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat