This product is now discontinued, View Related Products
RapidVap Vacuum Evaporation System
  • 1000-watt heater quickly reduces samples to dryness or an endpoint volume
  • Convenient, easy-to-set programming—store up to nine programs
  • Use a combination of gentle forces - vortex motion, heat, and vacuum - to process multiple samples
  • Set time, heat, and speed via keypad; programming feature allows you to alter parameters while a program is in progress
Vacuum Evaporation Systems reduce samples for agrochemistry, mycology testing, and environmental testing. Process up to 110 samples at once; microprocessor-controlled motion minimizes bumping and sample loss. Alarm sounds when time expires or when sample has almost reached dryness. Models 26854-10 and -15 feature a lid heater that eliminates condensation and improves visibility when evaporating high boiling point solvents. What’s included: three feet of ½"-ID vacuum hose, two hose clamps, lid receptacle, and a 6½-ft three-wire cord with plug. The 115 VAC models include a 20 A plug; 230 VAC models include a 15 A plug. Require evaporator block, vacuum pump, and sample tubes, order separately.
All units are available with an optional RS-232 link that allows you to set parameters via computer—please call our Application Specialists for more information.
$25,052.05 - $27,685.00CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Condenser Type Evaporation Flask Size Max Speed (rpm)
Availability Pricing
Labconco 7900001 Vacuum evaporation system, 230 VAC, 50/60 Hz, 3.5 A RK-26854-05 Mfr # 7900001
COMPARE
$25,052.05CAD / Each
Labconco 7900003 Vacuum evaporation system with lid heater, 230 VAC, 50/60 Hz, 3.5 A RK-26854-15 Mfr # 7900003
COMPARE
$27,685.00CAD / Each