Ralston Instruments Quick-Test Hose Assemblies
Reinforced microbore tubing assemblies for quick, low-volume, high-pressure systems—ideal for pressure instrumentation, hand pumps, and calibration sources/devices under test.
Feature brass connections.
$116.40 - $324.15CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Max Pressure (PSI) Description
Availability Pricing
Ralston Instruments QTHA-KIT6-3FT Quick-Test Hose Assembly Kit with Adapters; 3 ft RK-68976-75 Mfr # QTHA-KIT6-3FT
COMPARE
3 5000 Quick-Test Hose Assembly Kit with Adapters; 3 ft
$324.15CAD / Each
Ralston Instruments QTQT-HOS-3FT Quick-Test Hose Assembly; 3 ft RK-68976-92 Mfr # QTQT-HOS-3FT
COMPARE
3 6900 Quick-Test Hose Assembly; 3 ft
$116.40CAD / Each
Ralston Instruments QTQT-HOS-6FT Quick-Test Hose Assembly; 6 ft RK-68976-93 Mfr # QTQT-HOS-6FT
COMPARE
6 6900 Quick-Test Hose Assembly; 6 ft
$150.65CAD / Each
Ralston Instruments QTQT-HOS-10FT Quick-Test Hose Assembly; 10 ft RK-68976-94 Mfr # QTQT-HOS-10FT
COMPARE
10 6900 Quick-Test Hose Assembly; 10 ft
$194.12CAD / Each
Ralston Instruments QTQT-HOS-20FT Quick-Test Hose Assembly; 20 ft RK-68976-95 Mfr # QTQT-HOS-20FT
COMPARE
20 6900 Quick-Test Hose Assembly; 20 ft
$306.10CAD / Each