This product is now discontinued, View Related Products
RAE ToxiRAE 3 Single-Gas Monitors
Rugged stainless steel housing for harsh environments
  • Alarm conditions activate bright red flashing LEDS, loud audible alarm, and vibration alarm
Take this monitor wherever you need it — stainless steel housing is IP67-rated for water and dust resistance. Intuitive, easy-to-use user interface allows you to set high, low, STEL, and TWA alarms. Log up to ten events for later analysis. Secure your calibration settings, alarm limits, and data by password protecting the monitor.
Operate in display or countdown mode; display mode allows the unit to be programmed and turned on and off while countdown mode keeps the unit on until the sensor is depleted.
Approvals: CSA Class I, Div. 1, Groups A, B, C, D; CE; ATEX
$514.58 - $526.37CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
RAE Systems G01-0102-000 Single-Gas Detector, hydrogen sulfide (H2S) RK-86538-01 Mfr # G01-0102-000
COMPARE
Single-Gas Detector, hydrogen sulfide (H2S)
$514.58CAD / Each
RAE Systems G01-0101-000 Single-Gas Detector, carbon monoxide (CO), 0 to 500 ppm RK-86538-03 Mfr # G01-0101-000
COMPARE
Single-Gas Detector, carbon monoxide (CO), 0 to 500 ppm
$526.37CAD / Each
RAE Systems G01-0103-000 Single-Gas Detector, carbon monoxide (CO), 0 to 1999 ppm RK-86538-05 Mfr # G01-0103-000
COMPARE
Single-Gas Detector, carbon monoxide (CO), 0 to 1999 ppm
$523.05CAD / Each