Qsonica Q700 Touch-Screen Sonicator
Runs multiple programs in sequence
  • User-friendly touch screen control
  • Wide variety of applications — disrupt cells, shear DNA, mix compounds, extract samples, and nanoparticle dispersion
  • Self diagnostics and help screens simplify operation
This 700-watt ultrasonic processor tracks frequency changes in the convertor/tip assembly caused by load and temperature changes, and maintains optimal electrical efficiency at all times. Updated features include improved frequency and amplitude control, multiple sequence programming, and the ability to more accurately track the total energy delivered to your sample. The digital design allows the system to run more efficiently and requires less power to effectively process samples. This reduced energy consumption allows the unit to run cooler which extends the life of internal components. Pulse on/off from 1 second to 24 hours and total run time up to 72 hours. RoHS compliant.
$10,682.08 - $10,969.39CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (Hz) Power (VAC) Watts
Availability Pricing
Qsonica Q700 (120V) Touch-Screen Sonicator; 700 watt, 120VAC, 50/60 Hz RK-04712-82 Mfr # Q700 (120V)
COMPARE
50/60 120 700
$10,682.08CAD / Each
Qsonica Q700 (220V) Touch-Screen Sonicator; 700 watt, 220VAC, 50/60 Hz RK-04712-84 Mfr # Q700 (220V)
COMPARE
50/60 220 700
$10,969.39CAD / Each