PYREX® Brand 6404 Separatory Funnels
Squibb, pear-shaped, HDPE stopper, PTFE stopcock
  • Length of stem below stopcock is 65 mm
Stopper is manufactured with a closed bottom and is made from high-density polyethylene to conform to stopper dimensions.
$294.68 - $1,267.12CAD / Case of 1
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Stopcock Plug Size Stopper Number
Availability Pricing
Pyrex 6404-125 6404 Separatory Funnel, 125 mL, 1/pk RK-34507-32 Mfr # 6404-125
COMPARE
125 2 22
$294.68CAD / Each
Pyrex 6404-250 6404 Separatory Funnel, 250 mL, 1/pk RK-34507-34 Mfr # 6404-250
COMPARE
250 4 22
$375.71CAD / Each
Pyrex 6404-500 6404 Separatory Funnel, 500 mL, 1/pk RK-34507-36 Mfr # 6404-500
COMPARE
500 4 27
$427.28CAD / Each
Pyrex 6404-250 6404 Separatory Funnel, 250 mL, 4/cs RK-34507-54 Mfr # 6404-250
COMPARE
250 4 22
$1,105.04CAD / Case of 4
Pyrex 6404-500 6404 Separatory Funnel, 500 mL, 4/cs RK-34507-56 Mfr # 6404-500
COMPARE
500 4 27
$1,267.12CAD / Case of 4
Pyrex 6404-1L 6404 Separatory Funnel, 1000 mL, 1/cs RK-34507-58 Mfr # 6404-1L
COMPARE
1000 4 27
$456.75CAD / Case of 1
Pyrex 6404-2L 6404 Separatory Funnel, 2000 mL, 1/cs RK-34507-60 Mfr # 6404-2L
COMPARE
2000 6 38
$648.29CAD / Case of 1
Showing 1- 7 of 7