PYREX® Brand 5642 Class A Volumetric Flasks
No more broken stoppers
  • High-density polyethylene stopper won’t break if dropped
  • Volumetric flasks are constructed of borosilicate glass
These PYREX® Brand 5642 Class A volumetric flasks feature machine-blown bodies and heavy-beaded, heavy-tubing necks to provide excellent durability. Volumetric flasks include a linear high-density polyethylene stopper that conforms to stopper dimensions. The graduation line is sharp and permanent. Large white block letters make the inscriptions easy to read. The 2-mL flask is test tube shaped.
$67.78 - $1,281.85CAD / Pkg of 6
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Stopper Number Accepted Tolerance (mL)
Availability Pricing
Pyrex Brand 5642 Class A Volumetric Flask, 100 mL; 6/PK RK-34561-06 Mfr # 5642-100
COMPARE
100 13 ±0.08
$604.09CAD / Pkg of 6
Pyrex Brand 5642 Class A Volumetric Flask, 5 mL; 1/CS RK-34561-22 Mfr # 5642-5
COMPARE
5 8 ±0.02
$67.78CAD / Case of 1
Pyrex Brand 5642 Class A Volumetric Flask, 10 mL; 12/CS RK-34561-23 Mfr # 5642-10
COMPARE
10 9 ±0.02
$663.03CAD / Case of 12
Pyrex Brand 5642 Class A Volumetric Flask, 25 mL; 12/CS RK-34561-24 Mfr # 5642-25
COMPARE
25 9 ±0.03
$721.96CAD / Case of 12
Pyrex Brand 5642 Class A Volumetric Flask, 50 mL; 12/CS RK-34561-25 Mfr # 5642-50
COMPARE
50 9 ±0.05
$780.90CAD / Case of 12
Pyrex Brand 5642 Class A Volumetric Flask, 100 mL; 12/CS RK-34561-26 Mfr # 5642-100
COMPARE
100 13 ±0.08
$810.36CAD / Case of 12
Pyrex Brand 5642 Class A Volumetric Flask, 200 mL; 12/CS RK-34561-27 Mfr # 5642-200
COMPARE
200 13 ±0.10
$942.97CAD / Case of 12
Pyrex Brand 5642 Class A Volumetric Flask, 500 mL; 12/CS RK-34561-29 Mfr # 5642-500
COMPARE
500 19 ±0.2
$1,281.85CAD / Case of 12
Pyrex Brand 5642 Class A Volumetric Flask, 1000 mL; 6/CS RK-34561-30 Mfr # 5642-1L
COMPARE
1000 22 ±0.3
$788.26CAD / Case of 6
Pyrex Brand 5642 Class A Volumetric Flask, 2000 mL; 4/CS RK-34561-31 Mfr # 5642-2L
COMPARE
2000 27 ±0.5
$663.03CAD / Case of 4
Showing 1- 10 of 10