PYREX® Brand 5640 Class A Volumetric Flasks
Get accurate dilutions
  • Sharp, permanent graduation makes for easier diluting
  • Volumetric flasks are constructed of borosilicate glass
These PYREX® Brand 5640 Class A volumetric flasks feature machine-blown bodies for durability. Heavy-tubing neck accepts glass stoppers. Each flask has sharp, permanent graduation markings. Large white block letters make inscriptions easy to read. The 1-mL and 2-mL flasks are test-tube shaped. Sizes 3000 mL to 6000 mL have pennyhead stoppers.
$198.91 - $1,466.02CAD / Pkg of 1
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL)
Availability Pricing
Pyrex Brand 5640 Class A Volumetric Flask; 1000 mL; 1/PK RK-34505-11 Mfr # 5640-1L
COMPARE
1000
$198.91CAD / Pkg of 1
Pyrex Brand 5640 Class A Volumetric Flask; 1 mL; 12/CS RK-34505-21 Mfr # 5640-1
COMPARE
1
$766.16CAD / Case of 12
Pyrex Brand 5640 Class A Volumetric Flask; 2 mL; 12/CS RK-34505-22 Mfr # 5640-2
COMPARE
2
$817.73CAD / Case of 12
Pyrex Brand 5640 Class A Volumetric Flask; 5 mL; 12/CS RK-34505-23 Mfr # 5640-5
COMPARE
5
$648.29CAD / Case of 12
Pyrex Brand 5640 Class A Volumetric Flask; 10 mL; 12/CS RK-34505-24 Mfr # 5640-10
COMPARE
10
$766.16CAD / Case of 12
Pyrex Brand 5640 Class A Volumetric Flask; 50 mL; 12/CS RK-34505-26 Mfr # 5640-50
COMPARE
50
$891.40CAD / Case of 12
Pyrex Brand 5640 Class A Volumetric Flask; 100 mL; 12/CS RK-34505-27 Mfr # 5640-100
COMPARE
100
$950.34CAD / Case of 12
Pyrex Brand 5640 Class A Volumetric Flask; 200 mL; 12/CS RK-34505-28 Mfr # 5640-200
COMPARE
200
$1,090.31CAD / Case of 12
Pyrex Brand 5640 Class A Volumetric Flask; 250 mL; 12/CS RK-34505-29 Mfr # 5640-250
COMPARE
250
$1,178.71CAD / Case of 12
Pyrex Brand 5640 Class A Volumetric Flask; 500 mL; 12/CS RK-34505-30 Mfr # 5640-500
COMPARE
500
$1,466.02CAD / Case of 12
Pyrex Brand 5640 Class A Volumetric Flask; 1000 mL; 6/CS RK-34505-31 Mfr # 5640-1L
COMPARE
1000
$891.40CAD / Case of 6
Pyrex Brand 5640 Class A Volumetric Flask; 2000 mL; 4/CS RK-34505-32 Mfr # 5640-2L
COMPARE
2000
$751.43CAD / Case of 4
Pyrex Brand 5640 Class A Volumetric Flask; 3000 mL; 1/CS RK-34505-33 Mfr # 5640-3L
COMPARE
3000
$751.43CAD / Case of 1
Pyrex Brand 5640 Class A Volumetric Flask; 4000 mL; 1/CS RK-34505-34 Mfr # 5640-4L
COMPARE
4000
$847.20CAD / Case of 1
Pyrex Brand 5640 Class A Volumetric Flask; 5000 mL; 1/CS RK-34505-35 Mfr # 5640-5L
COMPARE
5000
$965.07CAD / Case of 1
Pyrex Brand 5640 Class A Volumetric Flask; 6000 mL; 1/CS RK-34505-36 Mfr # 5640-6L
COMPARE
6000
$1,134.51CAD / Case of 1
Showing 1- 16 of 16