Pyrex Brand 1396 Square Glass Media Bottles

These space-saving bottles are made of borosilicate glass for good chemical and thermal resistance. Bottles include matching polypropylene caps. Autoclavable.
$201.85 - $363.93CAD / Case of 10

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Overall Height (in)
Availability Pricing
Pyrex 1396-100 1396 Glass Media Bottle, Square, 100 mL, 10/cs RK-34594-22 Mfr # 1396-100
COMPARE
100 Glass 4 1/4
$201.85CAD / Case of 10
Pyrex 1396-250 1396 Glass Media Bottle, Square, 250 mL, 10/cs RK-34594-24 Mfr # 1396-250
COMPARE
250 Glass 5
$223.96CAD / Case of 10
Pyrex 1396-500 1396 Glass Media Bottle, Square, 500 mL, 10/cs RK-34594-26 Mfr # 1396-500
COMPARE
500 Glass 7
$284.36CAD / Case of 10
Pyrex 1396-1L 1396 Glass Media Bottle, Square, 1000 mL, 10/cs RK-34594-28 Mfr # 1396-1L
COMPARE
1000 Glass 8 3/4
$363.93CAD / Case of 10
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat