Pyrex Brand 1395 Media Bottles
Heavy-duty borosilicate glass—perfect for storage, mixing, and sampling
Bottles have permanent white enamel graduations and marking spots.
Caution: Do not tighten caps immediately after autoclaving.
$163.55 - $701.33CAD / Case of 0
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Pyrex 1395-25 Brand 1395 Media Storage Bottle w/ Screw Cap, 25 mL RK-34514-20 Mfr # 1395-25
COMPARE
25 5 Glass
$163.55CAD / Case of 10
Pyrex 1395-50 Brand 1395 Media Storage Bottle w/ Screw Cap, 50 mL RK-34514-21 Mfr # 1395-50
COMPARE
50 10 Glass
$163.55CAD / Case of 10
Pyrex 1395-100 Brand 1395 Media Storage Bottle w/ Screw Cap, 100 mL RK-34514-22 Mfr # 1395-100
COMPARE
100 20 Glass
$167.97CAD / Case of 10
Pyrex 1395-250 Brand 1395 Media Storage Bottle w/ Screw Cap, 250 mL RK-34514-23 Mfr # 1395-250
COMPARE
250 50 Glass
$185.65CAD / Case of 10
Pyrex 1395-500 Brand 1395 Media Storage Bottle w/ Screw Cap, 500 mL RK-34514-24 Mfr # 1395-500
COMPARE
500 100 Glass
$218.06CAD / Case of 10
Pyrex 1395-1L Brand 1395 Media Storage Bottle w/ Screw Cap, 1 L RK-34514-25 Mfr # 1395-1L
COMPARE
1000 100 Glass
$272.58CAD / Case of 10
Pyrex 1395-2L Media Storage Bottle w/ Screw Cap, 2 L; 10/Pk RK-34514-26 Mfr # 1395-2L
COMPARE
2000 200 Glass
$701.33CAD / Case of 10
Pyrex 1395-5L Brand 1395 Media Storage Bottle w/ Screw Cap, 5 L RK-34514-28 Mfr # 1395-5L
COMPARE
5000 500 Glass
$246.06CAD / Case of 1
Pyrex 1395-10L Brand 1395 Media Storage Bottle w/ Screw Cap, 10 L RK-34514-30 Mfr # 1395-10L
COMPARE
10000 1000 Glass
$377.19CAD / Case of 0
Showing 1- 9 of 9