This product is now discontinued, View Related Products
PURELAB Option-R Water Purification Systems
Systems produce laboratory-grade water from a potable supply
  • Microprocessor-controlled system management ensures consistent water quality
  • Intuitive, graphically displayed controls and easy-to-replace consumables provide user-friendly operation
  • Four models provide 10 to 15 MΩ-cm from 7 to 60 L/hr
Systems feature a long-life integral prefilter for removing chlorine and gross particulate contamination, a high-flux reverse osmosis membrane, a high-capacity purification pack for removing organic and inorganic ionic impurities, and a UV photo-oxidation cell to minimize bateria count. An integral recirculation pump maintains consistent water quality and inhibits microbial growth. Front-entry service doors provide easy access to consumables — making replacement quick and simple. Large LCD shows temperature-compensated water purity, production mode, reservoir level, and consumable life data. Safety features includes programmable audio-visual alarms, power fail-safe, and auto-restart. Smaller R-7 and R-15 models feature a height adjustable dispensing tap and RS-232 connectivity.
Reservoir required with system. Order separately.
$10,251.85 - $20,664.29CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Production (L/hour)
Availability Pricing
Elga Option-R 7 PURELAB Water Purification System RK-99266-50 Mfr # OR007XXM1
COMPARE
420
$10,251.85CAD / Each
Elga Option-R 15 PURELAB Water Purification System RK-99266-55 Mfr # OR015XXM1
COMPARE
900
$20,664.29CAD / Each
Elga OPT030PLXX11503 PURELAB Option-R 30 Water Purification System RK-99266-60 Mfr # OPT030PLXX11503
COMPARE
$12,793.45CAD / Each
Elga OPT060PLXX11504 PURELAB Option-R 60 Water Purification System RK-99266-65 Mfr # OPT060PLXX11504
COMPARE
$13,525.72CAD / Each