This product is now discontinued, View Related Products
PTFE Brushes
PTFE brushes are specifically designed for use in high temperature applications. They can withstand temperatures up to 500°F (260°C) and outlast Nylon, plastic and natural Boar bristles. Resistant to many caustic chemicals.
$61.51 - $63.72CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Handle Length (in) Length (in) Material
Availability Pricing
COMPARE
5 6 PTFE
$63.72CAD / Each
COMPARE
7.5 9 1/2 PTFE
$61.51CAD / Each