ProCal Primary Turbidity Standard Solutions
Instrument specific kits and standards ensure maximum accuracy
  • Safe, non-toxic formulations are traceable to Formazin
  • Two year shelf life
ProCal Primary Turbidity Standards are formulated to provide instrument-specific accuracy with a safe, non toxic formulation, and offer improvements in stability over polymer and formazin alternatives, two-year shelf life. U.S. EPA-approved to meet NPDES and NPDWR compliance reporting requirements.
$58.94 - $710.17CAD / Each
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
NTU Value
Availability Pricing
HF Scientific 52040 Hach ProCal Primary Standard Kit for Process Turbidimeter 1720 RK-99900-33 Mfr # 52040
COMPARE
0.0, 20
$710.17CAD / Pkg of 4
HF Scientific 52050 Hach ProCal Primary Standard for Process Turbidimeter 1720, 0.0 NTU RK-99900-34 Mfr # 52050
COMPARE
0.0
$96.51CAD / Each
HF Scientific 52070 Hach ProCal Primary Standard for Process Turbidimeter 1720, 1.0 NTU RK-99900-35 Mfr # 52070
COMPARE
1.0
$206.27CAD / Each
HF Scientific 52090 Hach ProCal Primary Standard for Process Turbidimeter 1720, 20 NTU RK-99900-36 Mfr # 52090
COMPARE
20
$181.23CAD / Each
HF Scientific 52150 Hach ProCal Primary Standard Kit for Laboratory Turbidimeter 2100N RK-99900-37 Mfr # 52150
COMPARE
0.0, 20, 200, 1000, 4000
$524.53CAD / Pkg of 1
HF Scientific 52170 Hach ProCal Primary Standard for Laboratory Turbidimeter 2100N, 0.0 NTU RK-99900-38 Mfr # 52170
COMPARE
0.0
$71.46CAD / Each
HF Scientific 52190 Hach ProCal Primary Standard for Laboratory Turbidimeter 2100N, 20 NTU RK-99900-39 Mfr # 52190
COMPARE
20
$117.13CAD / Each
HF Scientific 52210 Hach ProCal Primary Standard for Laboratory Turbidimeter 2100N, 200 NTU RK-99900-40 Mfr # 52210
COMPARE
200
$129.66CAD / Each
HF Scientific 52230 Hach ProCal Primary Standard for Laboratory Turbidimeter 2100N, 1000 NTU RK-99900-41 Mfr # 52230
COMPARE
1000
$131.13CAD / Each
HF Scientific 52400 Hach ProCal Primary Standard Kit for Portable Turbidimeter 2100P RK-99900-42 Mfr # 52400
COMPARE
0.0, 20, 100, 800
$431.70CAD / Pkg of 1
HF Scientific 52420 Hach ProCal Primary Standard for Portable Turbidimeter 2100P, 0.0 NTU RK-99900-43 Mfr # 52420
COMPARE
0.0
$71.46CAD / Each
HF Scientific 52440 Hach ProCal Primary Standard for Portable Turbidimeter 2100P, 20 NTU RK-99900-44 Mfr # 52440
COMPARE
20
$127.45CAD / Each
HF Scientific 52460 Hach ProCal Primary Standard for Portable Turbidimeter 2100P, 100 NTU RK-99900-45 Mfr # 52460
COMPARE
100
$139.24CAD / Each
HF Scientific 52480 Hach ProCal Primary Standard for Portable Turbidimeter 2100P, 800 NTU RK-99900-46 Mfr # 52480
COMPARE
800
$123.76CAD / Each
HF Scientific 52500 Hach ProCal Primary Standard Kit for Portable Turbidimeter 2100Q RK-99900-47 Mfr # 52500
COMPARE
0.0, 20, 100, 800
$384.55CAD / Pkg of 1
HF Scientific 52520 Hach ProCal Primary Standard for Portable Turbidimeter 2100Q, 0.0 NTU RK-99900-48 Mfr # 52520
COMPARE
0.0
$58.94CAD / Each
HF Scientific 52540 Hach ProCal Primary Standard for Portable Turbidimeter 2100Q, 20 NTU RK-99900-49 Mfr # 52540
COMPARE
20
$103.14CAD / Each
HF Scientific 52560 Hach ProCal Primary Standard for Portable Turbidimeter 2100Q, 100 NTU RK-99900-50 Mfr # 52560
COMPARE
100
$120.82CAD / Each
Showing 1- 18 of 18