This product is now discontinued, View Related Products
PRO Scientific SB-1 Silent Brushless Digital Homogenizer
Quiet and maintenance free
  • No need for ear protection—low decibel level protects your hearing
  • Years of trouble-free operation without particulates or sparking from carbon brushes
  • Bright digital display with variable speed makes adjusting for different materials easy and repeatable
This benchtop homogenizer runs clean and quiet, ideal for applications where contamination is a concern, such as biotech, pharmaceutical, and cleanroom use. Because the motor is brushless, a significant spark source is eliminated, so this homogenizer is suitable for processing volatile or flammable organic compounds.
Choose a generator probe based on the sample volume and dispersion type (order separately). The standard quick-connect generators are precision crafted from 316 stainless steel and PTFE. The flat-bottom probe is for small volumes and the saw-tooth creates enough shear to handle larger volumes and tough tissues.
$6,169.08CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
PRO Scientific SB-1 Silent Brushless Digital Homogenize with Stand; 120V RK-04751-28 Mfr # 01-01370
COMPARE
120
$6,169.08CAD / Each
PRO Scientific SB-1 Silent Brushless Digital Homogenize with Stand; 220V RK-04751-29 Mfr # 01-02370
COMPARE
220
$6,169.08CAD / Each