PrimeLock® Installation Tools Accessories
Installation tools require mandrel(s) and clamp insert(s) for operation. EPDM-lined clamp inserts eliminate surface marks on tubing following insert installation. Minimum tube union installations require the appropriately sized minimum tube union mandrel in addition to the standard installation mandrel. Directional sleeve mandrels prevent backwards installation of the insert, sizes 3/8" to 1-1/4".
$58.94 - $3,409.42CAD / Each
36 variations of this product are available.
See All Product Options Below
36 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Description
Availability Pricing
COMPARE
1 1/4 PFA Installation Mandrel, standard, 1.25" fittings; 1/ea
$279.94CAD / Each
COMPARE
3/4 PFA Installation Mandrel, directional sleeve, 3/4" fittings; 1/ea
$786.79CAD / Each
COMPARE
3/8 PFA Installation Mandrel, standard, 3/8" fittings; 1/ea
$241.64CAD / Each
COMPARE
PFA Installation Mandrel Kit, standard, 1/4"-1" fittings
$1,022.53CAD / Each
COMPARE
1 PFA Installation Mandrel, minimum tube union, 1" fittings; 1/ea
$125.97CAD / Each
COMPARE
3/4 PFA Installation Mandrel, minimum tube union, 3/4" fittings; 1/ea
$125.97CAD / Each
COMPARE
1 1/4 PFA Installation Mandrel, directional sleeve, 1.25" fittings; 1/ea
$682.18CAD / Each
COMPARE
1/2 PFA Installation Mandrel, minimum tube union, 1/2" fittings; 1/ea
$58.94CAD / Each
COMPARE
1/2 PFA Installation Mandrel, directional sleeve, 1/2" fittings; 1/ea
$798.58CAD / Each
COMPARE
PFA Installation Mandrel Kit, directional sleeve, 1/4"-1" fittings
$3,409.42CAD / Each
COMPARE
1 PFA Installation Mandrel, directional sleeve, 1" fittings; 1/ea
$739.64CAD / Each
COMPARE
EPDM-lined PFA Clamp Insert Kit, EPDM-lined PFA, 1/4" to 1" fittings
$1,706.19CAD / Each
COMPARE
3/4 PFA Clamp Insert, PFA, standard, 3/4" OD tubing; 2/set
$330.04CAD / Set of 2
COMPARE
1 PFA Clamp Insert, PFA, standard, 1" OD tubing; 2/set
$330.04CAD / Set of 2
COMPARE
1 1/4 PFA Clamp Insert, PFA, standard, 1-1/4" OD tubing; 2/set
$330.04CAD / Set of 2
COMPARE
1 1/4 EPDM-lined PFA Clamp Insert, EPDM-lined PFA, standard, 1-1/4" OD tubing; 2/set
$363.93CAD / Each
COMPARE
1 1/2 PFA Clamp Insert, PFA, standard, 1-1/2" OD tubing; 2/set
$492.11CAD / Set of 2
COMPARE
1 1/2 EPDM-lined PFA Clamp Insert, EPDM-lined PFA, standard, 1-1/2" OD tubing; 2/set
$601.14CAD / Each
COMPARE
1/2 PFA Clamp Insert, PFA, standard, 1/2" OD tubing; 2/set
$330.04CAD / Set of 2
COMPARE
1/2 EPDM-lined PFA Clamp Insert, EPDM-lined PFA, standard, 1/2" OD tubing; 2/set
$363.93CAD / Set of 2
Showing 1 - 20 of 36