This product is now discontinued, View Related Products
Precision Digital Loop Leader Series Loop-Powered Indicators
Designed for demanding process control applications
  • 5-digit backlit display with bargraph
  • Custom engineering units
  • HART® protocol transparent
The Loop Leader Series’ PD683 & PD688 were designed for demanding process control applications. Standard features include a full 5-digit backlit display with bargraph, on-screen custom engineering units, and max/min display capabilities. Square root and programmable exponent functions are standard features on these meters. With these features these indicators can display flow from a DP transmitter or open channel flow in weirs and flumes. Programming is easy through the front panel buttons, with no pots or jumpers to deal with, and can be scaled without a signal source. The Loop Leader Series meters go just about anywhere. The PD688, with FM, CSA and ATEX certifications, can be installed in just about any hazardous area.
$359.51 - $543.68CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
DIN Size Input Type
Availability Pricing
Precision Digital PD683-0K0 Loop-Powered Indicator, general purpose RK-61600-00 Mfr # PD683-0K0
COMPARE
1/8\"
$359.51CAD / Each
Precision Digital PD688-0K0 Loop-Powered Indicators, hazardous area RK-61600-01 Mfr # PD688-0K0
COMPARE
1/8\"
$543.68CAD / Each