This product is now discontinued, View Related Products
Powerblanket® Beacon TC Series Smart Temperature Controllers
Control and monitor temperatures remotely
  • Receive wireless alerts and notifications
  • Adjustable short cycle delay for refrigeration applications
  • Watertight—NEMA 4X, rugged, steel-enclosed casing
  • Backlit LCD makes for easy viewing
Compatible with a wide variety of heating or cooling devices, the Wi-Fi connected temperature controllers allow you to remotely monitor current conditions and adjust setpoints for your devices from a smartphone or computer using a web portal or mobile app. Receive wireless alerts and notifications when the temperature or other values go outside your defined thresholds. Alerts can be customized based on temperature, electrical current, voltage, and resistance readings. Easily log, access, and export historic temperature data for future reference or process analysis. Electrical current and voltage can also be collected, allowing you to analyze the duty cycle, energy usage, and operating cost of your cooling or heating equipment.
Local and remote access can be secured with user-specific PIN numbers. You can also assign multiple users login access with read-write or read-only permission levels.
$425.81 - $499.48CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Power (VAC) Description
Availability Pricing
Powerblanket Beacon TC-W1010 Smart Temperature Controller; 12 Amps RK-42025-52 Mfr # TCW1010
COMPARE
212 120 / 240 Smart Temperature Controller; 12 Amps
$440.54CAD / Each
Powerblanket Beacon TCW1110 Smart Temperature Controller; 16 Amps RK-42025-53 Mfr # TCW1110
COMPARE
212 120 / 240 Smart Temperature Controller; 16 Amps
$499.48CAD / Each
Powerblanket Beacon TC-W3010 Smart Temperature Controller; 24 Amps RK-42025-54 Mfr # TCW3010
COMPARE
212 120 Smart Temperature Controller; 24 Amps
$425.81CAD / Each