This product is now discontinued, View Related Products
Portable AC/DC Radiation Monitors
Versatile, easy-to-use monitors can be used in both laboratory and field applications
Rugged monitors are constructed with a NEMA-4X polycarbonate case that is corrosion resistant and splash-proof to withstand the rigors of every day use. Large, analog bar graph display is readable at a glance without complicated programming or electronics — just power up and go. Monitors operate on battery or AC power source for portable or stationary use. Model 86551-20 features a 2'' pancake probe to read alpha, beta, and gamma radiation — ideal for clothing, benchtop, or hand surface readings. Model 86551-22 features a 3'' scintillation probe to read gamma and X-ray radiation — suitable for entryway or area monitoring.
$3,933.95 - $5,058.15CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Range Sensor
Availability Pricing
Technical Associates LAM-10+P-15 Portable AC/DC Radiation Monitor with Pancake Probe RK-86551-20 Mfr # LAM-10+P-15
COMPARE
0.02 to 200 mR/hr; 0.1 to 10,000 cps Pancake probe
$3,933.95CAD / Each
Technical Associates LAM-10+PGS-31 Radiation monitor with scintillation probe RK-86551-22 Mfr # LAM-10+PGS-3L
COMPARE
0.03 to 30 mR/hr; 0.1 to 10,000 cps Scintillation probe
$5,058.15CAD / Each