Cole-Parmer® Essentials Polystyrene Weighing Dishes - Hexagonal Dishes (Anti-Static)
Hexagonal weigh dishes bend for easy pouring
  • Disposable
Translucent, anti-static, polystyrene; flexes easily for weighing, dispensing, or pouring liquids or solids. Unique hexagonal design for non-slip handling and pouring. Biologically inert; resistant to most dilute acids, aqueous solutions, and alcohol bases. Flat bottoms resist tipping. Hundreds of sampling and weighing applications.
$142.92 - $272.58CAD / Pkg of 500
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Diameter (in) Material
Availability Pricing
COMPARE
20 1 3/4 Polystyrene
$142.92CAD / Pkg of 500
COMPARE
50 3 Polystyrene
$160.60CAD / Pkg of 500
COMPARE
200 4 3/4 Polystyrene
$239.43CAD / Pkg of 500
COMPARE
350 5 1/2 Polystyrene
$272.58CAD / Pkg of 500