This product is now discontinued, View Related Products
PolyScience Histology Bath
Freeze samples quickly, preventing ice crystal development and sample distortion
  • Reaches minimum temperature in about one hour
  • Fast-freeze samples with continuous cooling to -76°F (-60°C)
  • Immerse samples easily using included basket and chuck holder
  • Perform maintenance quickly and efficiently via the front-mounted reservoir drain
Histology baths are ideal for freezing samples for enzyme studies. The stainless steel bath reservoir and cover resist corrosion and provide durability. This bath also features easy one button operation, and the DuraTop™ insulating top deck is comfortable to the touch during operation.
Note: Using flammable isopentane as the bath fluid, operation in a fume hood is advised. Please see our selection of Fume Hoods or contact our Application Specialists for assistance.
$5,143.60CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
PolyScience HBATH Histology Bath, -76F/-60C, 120 VAC RK-10123-00 Mfr # HBATH-60-A11B
COMPARE
115
$5,143.60CAD / Each
PolyScience HBATH Histology Bath, -76F/-60C, 240 VAC RK-10123-05 Mfr # HBATH-60-A12E
COMPARE
230
$5,143.60CAD / Each