This product is now discontinued, View Related Products
PolyScience Heating Circulating Baths
Extra-large backlit digital display can be easily seen from across a room
  • Waterproof control panel swivels 180°, allowing viewing at almost any angle
  • Unique LidDock® system stores the lid in open position for easy sample placement and drains condensation back into reservoir
PolyScience’s next-generation baths maintain precise temperature control for virtually any laboratory or process application. Advanced PID algorithms increase heat responsiveness, eliminate set point overshoot, and ensure a constant temperature. The unique WhisperCool™ environmental control system features part-load sound levels below that of conventional speech at a distance of two meters. The chemical resistant DuraTop™ top plate remains cool at high temperatures and is easily cleaned and disinfected. Variable-speed pump keeps the bath liquid at a uniform temperature. Rear inlet/outlet ports also allow use in external process applications.
$6,247.17CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Interface Power (VAC) Description
Availability Pricing
PolyScience AP Advanced Programmable 7L Heating Bath Circulator 120V/60Hz RK-12119-02 Mfr # AP07H200-A11B
COMPARE
USB-A & B, Ethernet, RS-232, RS-485, remote on/off, and external temperature probe 115 Performance Digital 7L Heating Bath Circulator 120V/60Hz
$6,247.17CAD / Each