Plas-Labs La Petite Glove Boxes
Minimize cross-contamination during sample manipulation
  • Easy to move and clean
  • Ready access to electric outlets in chamber
This compact glove box gives you an isolated, ergonomically correct work area for manipulating biological samples. The two-piece glove box is sealed with stainless steel clamps to form an air- and water-tight barrier. Operations are visible without distortion through angled acrylic front. Seal materials resist microbiological growth and rounded chamber construction makes thorough cleaning simple.
Two internal electrical outlets facilitate equipment use. Main chamber utilizes purging for gas exchange, while the transfer chamber can use vacuum/gas fill for quick exchanges through the 9.75" (24.8 cm) opening. The large door is 12" (30.5 cm) in diameter.
$15,380.72CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Chamber Depth (in) Chamber Height (in) Power (VAC)
Availability Pricing
Plas-Labs 815-PGB La Petite Glove Box, 31.5" Wide; 110 VAC RK-34750-20 Mfr # 815-PGB
COMPARE
25.5 21.5 110
$15,380.72CAD / Each