This product is now discontinued, View Related Products
Plas-Labs Hypoxia Chamber Glove Box
Control and monitor O2 and CO2 in chamber
This hypoxia chamber glove box is ideal for tissue culture work, including tumor cell and stem cell research. Large 8" (20 cm) touch screen lets you automatically control oxygen, temperature, and carbon dioxide levels within the chamber. Control panel is a passwordprotected, configurable PID that features one-touch on-screen datalogging, air circulation control, and can be configured for up to six individually controlled sampling chambers.
Transparent transfer chamber features a vacuum gauge and a white leveling tray for transferring liquids through the chamber. Glove box features O2 and CO2 sensors;
inert, N2, CO2, and O2 gas hookups; an Ethernet port for data logging; and a platinum RTD with sealing gland.
$40,474.02 - $40,709.77CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Plas-Labs 856-HYPO Hypoxia Glove Box Chamber, 41" Wide; 110 VAC RK-34755-00 Mfr # 856-HYPO
COMPARE
115
$40,474.02CAD / Each
Plas-Labs 856-HYPO/EXP Hypoxia Glove Box Chamber, 41" Wide; 220 VAC RK-34755-02 Mfr # 856-HYPO/EXP
COMPARE
115
$40,709.77CAD / Each