This product is now discontinued, View Related Products
Plas-Labs Controlled Atmosphere Glove Boxes
Ideal working conditions at a touch
  • Eliminate reactive air components or maintain perfect temperature/humidity
  • Fluorescent lighting and ergonomic design enable delicate detail work
Use these glove boxes to maintain ideal atmospheric conditions for manipulation and transferring of sensitive materials and organisms. Double-layered neoprene gaskets provide a tight seal (up to 1.5 mm Hg) to enable low-pressure vacuum/gas fill cycling to establish main chamber atmosphere. Vacuum-capable transfer chamber enables easy sample transfers without disturbing main chamber conditions. Clear acrylic top with built-in fluorescent light and white matched die-molded bottom illuminate your work area and reduce shadows.
$27,095.64 - $44,667.29CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Chamber Depth (in) Chamber Height (in) Power (VAC)
Availability Pricing
Plas-Labs 855-AC 110V Controlled atmosphere anaerobic glove box; 2 port; 110 VAC, 60 Hz RK-34750-10 Mfr # 855-AC 110V
COMPARE
28 26 110
$36,379.47CAD / Each
Plas-Labs 855-AC/EXP Controlled atmosphere anaerobic glove box; 2 port; 220 VAC, 50 Hz RK-34750-15 Mfr # 855-AC/EXP
COMPARE
28 26 220
$27,095.64CAD / Each
Plas-Labs 890-THC Temperature and Humidity Controlled glove box; 2 port; 110 VAC, 60 Hz RK-34750-40 Mfr # 890-THC
COMPARE
28 26 110
$43,089.29CAD / Each
Plas-Labs 890-THC/EXP Temperature and Humidity Controlled glove box; 2 port; 230 VAC, 50 Hz RK-34750-45 Mfr # 890-THC/EXP
COMPARE
28 26 110
$44,667.29CAD / Each