Pemtair Quick-Change Filtration Systems
Easy installation of complete filtration system with quick cartridge replacement
The quick-change filtration systems offer a complete solution in one simple design—several cartridges
will meet your water quality needs. Easily change the cartridge filter with a quarter-turn of the filter. The filter head features a built-in auto shut-off when cartridge is removed, eliminating the need for additional shutoff valves in your filter system. Connections on inlets and outlets are available with 1⁄4" NPT fittings, or 1⁄4" and 3⁄8" John Guest® push-to-connect fittings.
Each quick-change system includes a cartridge, filter head, and mounting bracket for use as a single unit; single cartridge systems are NSF certified. You can also combine cartridges in a series of up to three filters with the optional double and triple bracketed heads (sold separately).
$44.94 - $287.31CAD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Micron Rating Port Size
Availability Pricing
Pentair 155785-43 Quick-Change Filtration System Replacement Cartridge; 0.5um Carbon Block RK-01507-41 Mfr # 155785-43
COMPARE
0.5
$62.99CAD / Each
Pentair 155962-43 Quick-Change Filtration System Replacement Cartridge; 5um Carbon Block RK-01507-42 Mfr # 155962-43
COMPARE
5
$62.25CAD / Each
Pentair 155901-43 Quick-Change Filtration System Replacement Cartridge; 20um Activated Carbon RK-01507-43 Mfr # 155901-43
COMPARE
20
$44.94CAD / Each
Pentair 255706-43 Quick-Change Filtration System Replacement Cartridge; 50um Granular Carbon RK-01507-44 Mfr # 255706-43
COMPARE
50
$50.46CAD / Each
Pentair 255705-43 Quick-Change Filtration System Replacement Cartridge; 1um Carbon Block RK-01507-45 Mfr # 255705-43
COMPARE
1
$62.25CAD / Each
Pentair 257438-06 Quick-Change Filtration System Single-Head Bracket; 1/4" OD RK-01507-90 Mfr # 257438-06
COMPARE
$54.88CAD / Each
Pentair 257729-06 Quick-Change Filtration System Double-Head Bracket; 1/4" OD RK-01507-91 Mfr # 257729-06
COMPARE
$229.85CAD / Each
Pentair 257440-06 Quick-Change Filtration System Triple-Head Bracket; 1/4" OD RK-01507-93 Mfr # 257440-06
COMPARE
$285.84CAD / Each
Pentair 257441-06 Quick-Change Filtration System Triple-Head Bracket; 3/8" OD RK-01507-94 Mfr # 257441-06
COMPARE
$287.31CAD / Each
Showing 1- 9 of 9