Palmer Wahl Chart Recorder Paper

Replacement chart paper for your chart recorder
Compatible with 30018-series 8" Circular Chart Pressure Recorders.
$92.75 - $101.96CAD / Pkg of 100

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Palmer Wahl P-0-100-8 Chart Recorder Paper, 8" dia, 0-100 psi, 24-hour, 100/pk RK-30018-43 Mfr # P-0-100-8
COMPARE
Chart Recorder Paper, 8" dia, 0-100 psi, 24-hour, 100/pk
$101.96CAD / Pkg of 100
Palmer Wahl P-0-300-8 Chart Recorder Paper, 8" dia, 0-300 psi, 24-hour, 100/pk RK-30018-44 Mfr # P-0-300-8
COMPARE
Chart Recorder Paper, 8" dia, 0-300 psi, 24-hour, 100/pk
$92.75CAD / Pkg of 100
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat