Orochem OroSil™ C18 HPLC Columns
Compatible even with 100% aqueous mobile phase
  • Ideal for separation of polar, semi-polar and nonpolar compounds at low to medium pH
  • Specialized surface treatment provides high hydrophobicity
  • Suitable for C18 phase
These Orochem OroSil™ HPLC columns are uniquely designed for the separation of polar, semi-polar and nonpolar compounds at low to medium pH. With specialized surface treatment, the OroSil™ chemistry makes these columns compatible even with 100% aqueous mobile phase. Columns are known to be stable at pH 1.5-9.0.
$786.05 - $1,121.25CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Length (mm) Column Particle Size (µm) Phase
Availability Pricing
Orochem OroSil™ HPLC Column, C18, 3 µm, 150 mm L x 4.6 mm ID RK-06486-17 Mfr # O3FI-902
COMPARE
150 3 C18
$907.61CAD / Each
Orochem OroSil™ HPLC Column, C18, 5 µm, 150 mm L x 4.6 mm ID RK-06486-18 Mfr # O5FI-903
COMPARE
150 5 C18
$961.39CAD / Each
Orochem OroSil™ HPLC Column, C18, 3 µm, 250 mm L x 4.6 mm ID RK-06486-19 Mfr # O3HI-902
COMPARE
250 3 C18
$1,121.25CAD / Each
Orochem OroSil™ HPLC Column, C18, 5 µm, 250 mm L x 4.6 mm ID RK-06486-20 Mfr # O5HI-903
COMPARE
250 5 C18
$1,065.26CAD / Each
Orochem OroSil™ HPLC Column, C18, 5 µm, 50 mm L x 3.0 mm ID RK-06486-21 Mfr # O5BG-903
COMPARE
50 5 C18
$786.05CAD / Each