Orochem Gazelle™ C18 and C18-Polar UHPLC Columns
Increase throughput while lowering solvent consumption
  • Improve LC-MS results with inline process monitoring
  • Yield higher sensitivity and reproducibility in less run time
  • Suitable for C18 and C18-Polar phases
Orochem Gazelle™ UHPLC columns offer increased throughput, low solvent consumption, inline process monitoring, and improved LC-MS results. The silica type A manufacturing process yields a narrow particle size and pore size distribution, which gives you consistent and reproducible results. The hydrophobic spherical surface is also stable over a wide pH range (2-8) to allow for excellent method flexibility.
$1,097.68 - $1,189.03CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Length (mm) Phase
Availability Pricing
Orochem Gazelle™ UHPLC Column, C18, 1.7 µm, 30 mm L x 2.1 mm ID RK-06486-12 Mfr # G17AQ-931
COMPARE
30 C18
$1,097.68CAD / Each
Orochem Gazelle™ UHPLC Column, C18, 1.7 µm, 50 mm l x 2.1 mm ID RK-06486-13 Mfr # G17BQ-931
COMPARE
50 C18
$1,116.09CAD / Each
Orochem Gazelle™ UHPLC Column, C18-Polar, 1.7 µm, 100 mm L x 2.1 mm ID RK-06486-14 Mfr # G17DQ-880
COMPARE
100 C18-Polar
$1,189.03CAD / Each
Orochem Gazelle™ UHPLC Column, C18-Polar, 1.7 µm, 30 mm L x 2.1 mm ID RK-06486-15 Mfr # G17AQ-880
COMPARE
30 C18-Polar
$1,097.68CAD / Each