This product is now discontinued, View Related Products
Opto 22 SNAP PAC System - S Series and R Series
  • FEATURES - R Series
  • Convenient PAC package for small or distributed applications
  • Compact rack-mounted industrial controller with I/O processor included (PAC-R-series)
  • Two independent 10/100 Mbps Ethernet network interfaces
  • One RS-232 port with handshaking suitable for modern communications using PPP
  • Full-featured control and HMI software included with purchase.
The SNAP PAC System is an integrated system of hardware and software for industrial control remote monitoring and data acquisition applications
  • Standalone controller suitable for larger and more complex applications
  • Up to 32 ioControl flowcharts running simultaneously
  • Two 10/100 Mbps Ethernet interfaces for host and I/O communication over standard Ethernet networks
  • One RS-485 serial port for connection to Opto 22 Mistic I/O units
  • Two RS-232 serial ports (one with full handshaking) for PPP host
    connection or direct connections to serial devices
  • Multiple protocol support:TCP/IP SNMP SMTP FTP OptoMMP™
  • FTP server/client capability; includes file system
$0.03 - $1,760.70CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Input Type Max Voltage Input (VDC) Number of Channels
Availability Pricing
Opto 22 PACPROJECTBAS PAC Project Basic Software Suite RK-20011-77 Mfr # PACPROJECTBAS
COMPARE
$0.03CAD / Each
Opto 22 PACPROJECTPRO PAC Project Professional Software Suite RK-20011-78 Mfr # PACPROJECTPRO
COMPARE
$1,471.92CAD / Each
Opto 22 SNAP-AIV-32 SNAP 32-Channel AIV Analog Input Module RK-20011-92 Mfr # SNAP-AIV-32
COMPARE
Volts 32
$1,466.02CAD / Each
Opto 22 PACPROJECTPRO PAC Contol Professional Software Suite RK-45000-03 Mfr # PACCONTROLPRO
COMPARE
$587.88CAD / Each
Opto 22 PACPROJECTPRO PAC Display Professional Software Suite RK-45000-04 Mfr # PACDISPLAYPRO
COMPARE
$587.88CAD / Each
Opto 22 SNAP-PAC-S1 Snap-pac-s1 Opto Controller RK-68473-50 Mfr # SNAP-PAC-S1
COMPARE
$1,760.70CAD / Each