This product is now discontinued, View Related Products
Opto 22 SNAP Brains
SNAP brains are I/O and communications processors that control analog, digital, or special-purpose I/O modules on the same compact SNAP B series mounting rack. SNAP Ethernet brains are compact, flexible, high-performance processors that connect real-world analog, digital, and serial devices with Ethernet networks for monitoring, control, and data acquisition. The SNAP-B3000-ENET works with analog, digital, and serial signals from real-world devices, while the SNAP-ENET-D64 Ethernet brain handles digital signals only. Both SNAP Ethernet brains feature pure TCP/IP transport protocol; a 10/100 Mbps Fast Ethernet connection; and multiple simultaneous protocols including Modbus/TCP, FTP, SNMP, and SMTP. Versions of the SNAP-B3000-ENET with a built-in realtime clock or 802.11 wireless LAN connectivity are also available.
Serial brains support both Optomux and Mistic protocols for easy integration into existing Optomux or Mistic installations. SNAP serial brains offer a variety of high-speed serial communications options and can be daisy-chained on a simple serial link over very long distances. The Mistic protocol provides seamless integration with Opto 22’s FactoryFloor software.
$0.03 - $1,471.92CAD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Input Type Max Voltage Input (VDC) Number of Channels
Availability Pricing
Opto 22 SNAP-PAC-EB1 PAC Remote I/O, High Density Speed Count RK-20012-23 Mfr # SNAP-PAC-EB1
COMPARE
$950.34CAD / Each
Opto 22 SNAP-PAC-EB2 Remote I/O Brain Module RK-68473-51 Mfr # SNAP-PAC-EB2
COMPARE
Digital, Analog, Serial
$663.03CAD / Each
Opto 22 SNAP-B3000-MODBUS B3000:Modbus Snap Analog/Digital Slave Brain RK-68474-66 Mfr # SNAP-B3000-MODBUS
COMPARE
$699.86CAD / Each
Opto 22 SNAP-B3000-ENET Opto Snap B3000ENET: Analog/Digital/Serial Ethernet Brain RK-68475-14 Mfr # SNAP-B3000-ENET
COMPARE
$1,141.88CAD / Each
Opto 22 SNAP-BRS Opto Snap Brs: Remote Digital Read/Write Brain, Mistic Protocol, Serial RK-68475-17 Mfr # SNAP-BRS
COMPARE
$478.85CAD / Each
Opto 22 SNAP-ENET-D64 Snap-Enet-D64:Digital Only Ethernet Brain RK-68475-19 Mfr # SNAP-ENET-D64
COMPARE
$1,141.88CAD / Each
Opto 22 SNAP-ENET-RTC Snap-Enet-Rtc Analog/Digital/Serial Ethernet Brain With Real Time Clock RK-68475-20 Mfr # SNAP-ENET-RTC
COMPARE
$1,215.55CAD / Each
Opto 22 SNAP-PDPRS64 Profibus-DP Digital Slave Brain RK-68475-49 Mfr # SNAP-PDPRS64
COMPARE
$817.73CAD / Each
Opto 22 SNAP-WLAN-FH-ADS Snap-Wlan-Fh-Ads Analog/Digital/ Serial Wireless Lan Ethernet Brain RK-68475-61 Mfr # SNAP-WLAN-FH-ADS
COMPARE
$0.03CAD / Each
Opto 22 B3000-B Analog Digital Brain; Mistic/B-Version RK-68475-73 Mfr # B3000-B
COMPARE
Digital, Analog, Serial
$1,105.04CAD / Each
Opto 22 SNAP-PAC-EB1 Serial Brain, Analog/Digital/Serial RK-68493-57 Mfr # SNAP-PAC-SB2
COMPARE
Digital, Analog, Serial
$736.70CAD / Each
Opto 22 SNAP-PAC-EB1 Wireless SNAP PAC Ethernet Brain, Ana/Dig/Ser with High-speed Digital Counting RK-68493-58 Mfr # SNAP-PAC-EB1-W
COMPARE
Digital, Analog, Serial
$1,471.92CAD / Each
Showing 1- 12 of 12