This product is now discontinued, View Related Products
Opto 22 Printed Circuit Series Solid State Relays
The MP Series packaging is designed with a minimum footprint to allow maximum relay density on the circuit board. All MP Series SSRs provide 4000 VRMS of optical isolation and provide a wide operating temperature range of -40°C to +100°C. MP Series SSRs are TTL compatible, with a 3-32 VDC control voltage. All MP Series SSRs feature a built-in snubber and a maximum zero voltage turn-on time of 1/2 cycle. All models are UL recognized and CSA certified.
Opto 22’s Printed Circuit Series allows OEMs to easily deploy solid-state relays on printed circuit boards with switch loads up to 4 amps.
$20.63 - $32.41CAD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Input Power (VAC) Max Current (amps)
Availability Pricing
Opto 22 DC200MP DC Control SSR, 200 VDC, 1 A RK-68474-26 Mfr # DC200MP
COMPARE
1
$29.47CAD / Each
Opto 22 DC60MP DC Control SSR, 60 VDC, 3 A RK-68474-27 Mfr # DC60MP
COMPARE
3
$20.63CAD / Each
Opto 22 MP120D2 DC Control SSR, 120 VAC, 2 A RK-68474-28 Mfr # MP120D2
COMPARE
2
$21.73CAD / Each
Opto 22 MP120D4 DC Control SSR, 120 VAC, 4 A RK-68474-29 Mfr # MP120D4
COMPARE
4
$27.99CAD / Each
Opto 22 MP240D2 DC Control SSR, 240 VAC, 2 A RK-68474-30 Mfr # MP240D2
COMPARE
2
$24.31CAD / Each
Opto 22 MP240D4 DC Control SSR, 240 VAC, 4 A RK-68474-31 Mfr # MP240D4
COMPARE
4
$32.41CAD / Each
Opto 22 MP380D4 DC Control SSR, 380 VAC, 4 A RK-68474-32 Mfr # MP380D4
COMPARE
4
$27.99CAD / Each
Opto 22 MP380D4 Solid State Relay (SSR); 240VAC/4A/DC Control RK-68494-27 Mfr # MP240D4-17
COMPARE
4
$32.41CAD / Each
Opto 22 DC60MP 120 VAC, 2 Amp, DC Control Solid State Relay (SSR) RK-68494-29 Mfr # P120D2
COMPARE
3
$25.56CAD / Each
Opto 22 DC60MP 120 VAC, 4 Amp, DC Control Solid State Relay (SSR) RK-68494-30 Mfr # P120D4
COMPARE
3
$27.11CAD / Each
Opto 22 DC60MP 240VAC, 2 Amp, DC Control Solid State Relay (SSR) RK-68494-31 Mfr # P240D2
COMPARE
3
$27.11CAD / Each
Opto 22 DC60MP 240VAC, 4 Amp, DC Control Solid State Relay (SSR) RK-68494-32 Mfr # P240D4
COMPARE
3
$27.99CAD / Each
Showing 1- 12 of 12