This product is now discontinued, View Related Products

Opto 22 Mistic-Protocol IO Bricks

Opto 22 Mistic-protocol I/O units are a family of intelligent digital and analog input/output controllers that operate as slave devices to a host computer. Physically, each Mistic I/O unit is a modular assembly that consists of a module mounting rack, a brain board, and a number of optically-isolated analog or digital I/O modules.
$1,024.01 - $1,225.86CAD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Input Type Max Voltage Input (VDC) Number of Channels
Availability Pricing
Opto 22 G4PS245B Mistic Classic Controller Power Supply + 7 Bricks 220 Vac RK-68474-75 Mfr # G4PS245B
COMPARE
$1,024.01CAD / Each
Opto 22 G4A8R Opto Mistic :8PT Remote Mf Brick RK-68475-78 Mfr # G4A8R
COMPARE
$1,178.71CAD / Each
Opto 22 G4D16R Multifunction I/O Unit, Mistic protocol, 16-channel RK-68475-80 Mfr # G4D16R
COMPARE
Digital
$1,225.86CAD / Each
Opto 22 G4PS245A Classic Controller Power Supply, 120 VAC, +7 bricks RK-68475-82 Mfr # G4PS245A
COMPARE
$1,099.15CAD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat