Diba Omnifit® EZ SolventPlus™ Chromatography Columns with One Fixed and One Adjustable Endpieces
PTFE, borosilicate glass, and Chemraz® wetted materials offer exceptional chemical resistance
  • PEEK™ column threads offer outstanding durability
  • Ergonomically designed adjustable endpiece offers quick and easy height adjustment
  • Superior frit placement adds bed support and even distribution of sample flow
  • Reusable columns let you pack your own media specific to your application
Use these EZ SolventPlus™ columns for techniques using organic solvents such as reversed phase chromatography. EZ SolventPlus columns have only the most chemically inert materials in the fluid path—borosilicate glass, PTFE, and Chemraz®—ensuring the utmost chemical resistance. Columns are ideal for low to medium pressure applications and are resistant to many solvents used in liquid chromatography. Modular design gives you the flexibility to mix and match column sizes with different endpiece types to meet your specific requirements.
One fixed endpiece is used as a bed support and one adjustable endpiece (plunger) lets you increase the bed height up to 80 mm, plus gives you the ability to make fine adjustments with minimal disturbance to the packed bed. Each glass column assembly is supplied preassembled with 30 μm PTFE frits and comes with all the tubing, fittings, and connections you need to get started.
$957.70 - $3,720.31CAD / Each
36 variations of this product are available.
See All Product Options Below
36 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Max Pressure (PSI)
Availability Pricing
Diba Omnifit® EZ SolventPlus™ Chromatography Column with 1 Fixed and 1 Adjustable Endpiece, 6.6 mm ID x 50 mm L RK-11943-87 Mfr # 006EZS-06-05-AF
COMPARE
6.6 50 900
$957.70CAD / Each
Diba Omnifit® EZ SolventPlus™ Chromatography Column with 1 Fixed and 1 Adjustable Endpiece, 6.6 mm ID x 100 mm L RK-11943-90 Mfr # 006EZS-06-10-AF
COMPARE
6.6 100 900
$1,068.21CAD / Each
Diba Omnifit® EZ SolventPlus™ Chromatography Column with 1 Fixed and 1 Adjustable Endpiece, 6.6 mm ID x 150 mm L RK-11943-93 Mfr # 006EZS-06-15-AF
COMPARE
6.6 150 900
$979.80CAD / Each
Diba Omnifit® EZ SolventPlus™ Chromatography Column with 1 Fixed and 1 Adjustable Endpiece, 6.6 mm ID x 250 mm L RK-11943-96 Mfr # 006EZS-06-25-AF
COMPARE
6.6 250 900
$987.17CAD / Each
Diba Omnifit® EZ SolventPlus™ Chromatography Column with 1 Fixed and 1 Adjustable Endpiece, 6.6 mm ID x 330 mm L RK-11943-99 Mfr # 006EZS-06-33-AF
COMPARE
6.6 330 900
$1,038.74CAD / Each
Diba Omnifit® EZ SolventPlus™ Chromatography Column with 1 Fixed and 1 Adjustable Endpiece, 6.6 mm ID x 400 mm L RK-11944-02 Mfr # 006EZS-06-40-AF
COMPARE
6.6 400 900
$1,038.74CAD / Each
Diba Omnifit® EZ SolventPlus™ Chromatography Column with 1 Fixed and 1 Adjustable Endpiece, 10 mm ID x 100 mm L RK-11944-05 Mfr # 006EZS-10-10-AF
COMPARE
10 100 600
$1,237.65CAD / Each
Diba Omnifit® EZ SolventPlus™ Chromatography Column with 1 Fixed and 1 Adjustable Endpiece, 10 mm ID x 150 mm L RK-11944-08 Mfr # 006EZS-10-15-AF
COMPARE
10 150 600
$1,237.65CAD / Each
Diba Omnifit® EZ SolventPlus™ Chromatography Column with 1 Fixed and 1 Adjustable Endpiece, 10 mm ID x 250 mm L RK-11944-11 Mfr # 006EZS-10-25-AF
COMPARE
10 250 600
$1,252.38CAD / Each
Diba Omnifit® EZ SolventPlus™ Chromatography Column with 1 Fixed and 1 Adjustable Endpiece, 10 mm ID x 330 mm L RK-11944-14 Mfr # 006EZS-10-33-AF
COMPARE
10 330 600
$1,267.12CAD / Each
Diba Omnifit® EZ SolventPlus™ Chromatography Column with 1 Fixed and 1 Adjustable Endpiece, 10 mm ID x 400 mm L RK-11944-17 Mfr # 006EZS-10-40-AF
COMPARE
10 400 600
$1,281.85CAD / Each
Diba Omnifit® EZ SolventPlus™ Chromatography Column with 1 Fixed and 1 Adjustable Endpiece, 10 mm ID x 500 mm L RK-11944-20 Mfr # 006EZS-10-50-AF
COMPARE
10 500 600
$1,333.42CAD / Each
Diba Omnifit® EZ SolventPlus™ Chromatography Column with 1 Fixed and 1 Adjustable Endpiece, 15 mm ID x 100 mm L RK-11944-23 Mfr # 006EZS-15-10-AF
COMPARE
15 100 300
$1,458.66CAD / Each
Diba Omnifit® EZ SolventPlus™ Chromatography Column with 1 Fixed and 1 Adjustable Endpiece, 15 mm ID x 150 mm L RK-11944-26 Mfr # 006EZS-15-15-AF
COMPARE
15 150 300
$1,451.29CAD / Each
Diba Omnifit® EZ SolventPlus™ Chromatography Column with 1 Fixed and 1 Adjustable Endpiece, 15 mm ID x 250 mm L RK-11944-29 Mfr # 006EZS-15-25-AF
COMPARE
15 250 300
$1,407.09CAD / Each
Diba Omnifit® EZ SolventPlus™ Chromatography Column with 1 Fixed and 1 Adjustable Endpiece, 15 mm ID x 330 mm L RK-11944-32 Mfr # 006EZS-15-33-AF
COMPARE
15 330 300
$1,466.02CAD / Each
Diba Omnifit® EZ SolventPlus™ Chromatography Column with 1 Fixed and 1 Adjustable Endpiece, 15 mm ID x 400 mm L RK-11944-35 Mfr # 006EZS-15-40-AF
COMPARE
15 400 300
$1,429.19CAD / Each
Diba Omnifit® EZ SolventPlus™ Chromatography Column with 1 Fixed and 1 Adjustable Endpiece, 15 mm ID x 500 mm L RK-11944-38 Mfr # 006EZS-15-50-AF
COMPARE
15 500 300
$1,458.66CAD / Each
Diba Omnifit® EZ SolventPlus™ Chromatography Column with 1 Fixed and 1 Adjustable Endpiece, 25 mm ID x 100 mm L RK-11944-41 Mfr # 006EZS-25-10-AF
COMPARE
25 100 150
$1,606.00CAD / Each
Diba Omnifit® EZ SolventPlus™ Chromatography Column with 1 Fixed and 1 Adjustable Endpiece, 25 mm ID x 150 mm L RK-11944-44 Mfr # 006EZS-25-15-AF
COMPARE
25 150 150
$1,628.10CAD / Each
Showing 1 - 20 of 36