This product is now discontinued, View Related Products
Omegadyne Tension and Compression Load Cells - Sub-miniature
Sub-miniature load cell
Small in size and capable of highly accurate readings, the LCFD Series comprises subminiature load cells for precision industrial applications. They are all stainless steel, measure both tension and compression
loads, and have male thread studs for load attachment. The exclusive internal design provides superior long-term stability and minimizes the effects of small off-axis loads.
$1,212.60 - $1,670.82CAD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Capacity (lbs)
Availability Pricing
Omegadyne LCFD-1K Load Cell, 1000 lb capacity, Sub-Miniature Load Cell RK-59890-42 Mfr # LCFD-1K
COMPARE
1000
$1,670.82CAD / Each
Omegadyne LCFD-10 Load Cell 10LB Capacity Sub-Miniature Load Cell RK-93955-55 Mfr # LCFD-10
COMPARE
10
$1,212.60CAD / Each
Omegadyne LCFD-100 Load Cell 100LB Capacity Sub-Miniature Load Cell RK-93955-56 Mfr # LCFD-100
COMPARE
100
$1,609.68CAD / Each
Omegadyne LCFD-1KG Load Cell 1000LB Capacity Sub-Miniature Load Cell RK-93955-57 Mfr # LCFD-1KG
COMPARE
2.2
$1,609.68CAD / Each
Omegadyne LCFD-25 Load Cell 25LB Capacity Sub-Miniature Load Cell RK-93955-58 Mfr # LCFD-25
COMPARE
25
$1,617.05CAD / Each
Omegadyne LCFD-250 Load Cell 250LB Capacity Sub-Miniature Load Cell RK-93955-59 Mfr # LCFD-250
COMPARE
250
$1,609.68CAD / Each
Omegadyne LCFD-5 Load Cell 5LB Capacity Sub-Miniature Load Cell RK-93955-60 Mfr # LCFD-5
COMPARE
5
$1,609.68CAD / Each
Omegadyne LCFD-50 Load Cell 50LB Capacity Sub-Miniature Load Cell RK-93955-61 Mfr # LCFD-50
COMPARE
50
$1,609.68CAD / Each
Omegadyne LCFD-500 Load Cell 500LB Capacity Sub-Miniature Load Cell RK-93955-62 Mfr # LCFD-500
COMPARE
500
$1,609.68CAD / Each
Showing 1- 9 of 9