This product is now discontinued, View Related Products
Omegadyne Industrial Load Cells - Sub-Miniature
Sub-miniature, very low profileThese sub-miniature compression load cells are designed to measure load ranges from 1 kg to 1000 lb. Rugged all stainless steeel construction and high-performance strain gauges assure superior linearity and stability. These cells are designed to be mounted on a smooth, flat surface.
$969.49 - $1,182.03CAD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Capacity (lbs)
Availability Pricing
Newport LCKD-1KG Sub-Miniature Load Cell; 1 kg (2.2 lb) RK-59887-89 Mfr # LCKD-1KG/N
COMPARE
2.2
$1,043.16CAD / Each
Newport LCKD-250 Sub-Miniature Load Cell; 250 lb RK-59887-91 Mfr # LCKD-250/N
COMPARE
250
$985.70CAD / Each
Newport LCKD-5 Sub-Miniature Load Cell; 5 lb RK-59887-92 Mfr # DISCONTINUED
COMPARE
5
$994.54CAD / Each
Newport LCKD-10 Sub-Miniature Load Cell; 10 lb RK-59890-89 Mfr # LCKD-10/N
COMPARE
10
$969.49CAD / Each
Newport LCKD-100 Sub-Miniature Load Cell; 100 lb RK-59890-90 Mfr # LCKD-100/N
COMPARE
100
$985.70CAD / Each
Newport LCKD-25 Sub-Miniature Load Cell; 25 lb RK-59890-92 Mfr # LCKD-25/N
COMPARE
25
$975.38CAD / Each
Newport LCKD-50 Sub-Miniature Load Cell; 50 lb RK-59890-95 Mfr # LCKD-50/N
COMPARE
50
$985.70CAD / Each
Newport LCKD-500 Sub-Miniature Load Cell; 500 lb RK-59890-96 Mfr # LCKD-500/N
COMPARE
500
$969.49CAD / Each
Omegadyne LCKD1000 Subminiature Compression Load Cells, 1000 LB capacity RK-59890-97 Mfr # LCKD1000
COMPARE
1000
$1,182.03CAD / Each
Showing 1- 9 of 9