Cole-Parmer® Essentials Single-Channel Fixed-Volume Pipettor
Ergonomic finger grip for comfortable use
  • Easy to operate with its ergonomic, lightweight push-button design
  • Forward and reverse pipetting modes
  • Drop of the pipette is color-coded by volume for easy identification
  • Volume is displayed through handgrip window for quick glance convenience
These pipettors are offered in a wide-range of fixed volumes to lend to a variety of precise liquid dispensing needs. They are fully autoclavable.
$54.07 - $129.66CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy (%) Color Code Max Volume (µL)
Availability Pricing
COMPARE
±0.8 Yes 10
$54.07CAD / Each
COMPARE
1.3 Yellow 5
$128.18CAD / Each
COMPARE
0.5 Yellow 50
$116.40CAD / Each
COMPARE
0.5 Yellow 100
$116.40CAD / Each
COMPARE
0.3 Blue 500
$116.40CAD / Each
COMPARE
0.3 Blue 1000
$129.66CAD / Each
COMPARE
0.3 Green 5000
$116.40CAD / Each
COMPARE
0.6 Green 10000
$116.40CAD / Each
Showing 1- 8 of 8