Cole-Parmer® Single and Multichannel Electronic Pipettors
Ergonomic design for effortless dispensing
  • Five essential modes: forward, reverse, multi, mixing and diluting
  • 360° rotational manifold for pipetting convenience
  • Auto-sensing magnetic piston for controlled pipetting of individual channels
  • Specially designed tip cones for leak-free operation
  • Drop of the pipette is color-coded by volume for easy identification
  • Smooth and light-loading ejection of tips
Designed to operate up to 5 different modes of pipetting, these pipettes also feature the capability of storing up to three of your more frequently used modes. Another feature is a display panel for ease of use. They have a Smart Power Saver, which activates a sleep mode enhancing its battery life.
Its battery charges by either a charging stand or USB power cord and does so in two hours, lasting up to 5 hours of continuous pipetting.
$673.34 - $1,370.25CAD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Code Max Volume (µL) Number Of Channels
Availability Pricing
COMPARE
Orange 10 1
$673.34CAD / Each
COMPARE
Magenta 100 1
$673.34CAD / Each
COMPARE
Yellow 200 1
$673.34CAD / Each
COMPARE
Blue 1000 1
$673.34CAD / Each
COMPARE
Green 5000 1
$673.34CAD / Each
COMPARE
Orange 10 8
$1,205.23CAD / Each
COMPARE
Green 20 8
$1,205.23CAD / Each
COMPARE
Magenta 100 8
$1,205.23CAD / Each
COMPARE
Yellow 200 8
$1,205.23CAD / Each
COMPARE
Blue 300 8
$1,205.23CAD / Each
COMPARE
Orange 10 12
$1,370.25CAD / Each
COMPARE
Green 20 12
$1,370.25CAD / Each
COMPARE
Magenta 100 12
$1,370.25CAD / Each
COMPARE
Yellow 200 12
$1,370.25CAD / Each
COMPARE
Blue 300 12
$1,370.25CAD / Each
Showing 1- 15 of 15