This product is now discontinued, View Related Products
Ohaus Pioneer PA+ Analytical Balances
Great for basic routine weighing in a variety of laboratory, industrial, and education applications
  • Bright backlit LCD is easy to view in any environment
  • Durable construction with metal base, ABS top housing, and stainless steel pan
  • Adjustable zero tracking is ideal for slow-filling applications
  • Adjustable balance sensitivity to handle environmental disturbances or application requirements
  • An up-front level indicator eliminates the need to look behind the balance during leveling
The Pioneer’s draft shield is designed with all glass panels including three sliding doors. All panels are easy to remove and replace, along with all components making it fast and easy to clean within seconds. Periodic cleaning helps avoid sample contamination and extends the life of the balance. A standard in-use cover provides added protection against spillage and minimizes damage to the display and keypad.
Three user-selectable environmental settings help the balance adapt to harsh environments and other disturbances for greater weighing accuracy. An RS-232 interface allows connection to a printer or PC for a record of balance ID, user and project ID, and time and date. Other features include backlight display, level indicator, and menu lock.
$1,547.06 - $3,860.28CAD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Calibration Type Capacity (g) Readability (g)
Availability Pricing
Ohaus PA84 Pioneer Analytical Balance 85 g x 0.0001 g RK-11611-40 Mfr # PA84
COMPARE
External 85 0.0001
$2,622.63CAD / Each
Ohaus PA124 Pioneer Analytical Balance 120 g x 0.0001 g RK-11611-41 Mfr # PA124
COMPARE
External 120 0.0001
$3,123.59CAD / Each
Ohaus PA224 Pioneer Analytical Balance 220 g x 0.0001 g RK-11611-42 Mfr # PA224
COMPARE
External 220 0.0001
$3,462.47CAD / Each
Ohaus PA163 Pioneer Toploading Balance 160g x 0.001g RK-11611-43 Mfr # PA163
COMPARE
External 160 0.001
$1,547.06CAD / Each
Ohaus PA323 Pioneer Toploading Balance 320g x 0.001g RK-11611-44 Mfr # PA323
COMPARE
External 320 0.001
$1,827.00CAD / Each
Ohaus PA523 Pioneer Toploading Balance 520g x 0.001g RK-11611-45 Mfr # PA523
COMPARE
External 520 0.01
$2,210.09CAD / Each
Ohaus PA84C Pioneer Analytical Balance 85 g x 0.0001 g with Internal Calibration RK-11611-52 Mfr # PX84
COMPARE
Internal 85 0.0001
$2,968.88CAD / Each
Ohaus PA124C Pioneer Analytical Balance 120 g x 0.0001 g with Internal Calibration RK-11611-53 Mfr # PA124C
COMPARE
Internal 120 0.0001
$3,506.67CAD / Each
Ohaus PA224C Pioneer Analytical Balance 220 g x 0.0001 g with Internal Calibration RK-11611-54 Mfr # PA224C
COMPARE
Internal 220 0.0001
$3,860.28CAD / Each
Ohaus PA163C Pioneer Toploading Balance 160g x 0.001g with Internal Calibration RK-11611-55 Mfr # PA163C
COMPARE
Internal 160 0.001
$1,900.67CAD / Each
Ohaus PA323C Pioneer Toploading Balance 320g x 0.001g with Internal Calibration RK-11611-56 Mfr # PA323C
COMPARE
Internal 320 0.001
$2,165.88CAD / Each
Ohaus PA523C Pioneer Toploading Balance 520g x 0.001g with Internal Calibration RK-11611-57 Mfr # PA523C
COMPARE
Internal 520 0.01
$2,541.60CAD / Each
Showing 1- 12 of 12