This product is now discontinued, View Related Products
Ohaus CS Portable Compact Scales
Compact, portable scales are ideal for use in light industrial, field, or office use
  • RF protected for use in production areas
  • Digital keypad calibration with external weight
  • Overload and underload indicators
The OHAUS Compact Scale is a lightweight, portable scale perfectly suited for use in laboratory, industrial, education and home use applications. Applications can include forensic testing, quality control, formulation, soil sampling, postal weighing and dietary needs.
Scales feature a large weighing platform and can adequately handle large samples including oversized letters. In addition, the standard stainless steel platform is removable for easy cleaning.
The low profile design is truly portable when powered by 3 “AA” batteries (included). A standard AC adapter can also be used to power the scales when used in a more permanent location.
These scales offer easy operation with 2 clearly marked buttons, low battery indicator and auto shut-off to conserve battery life. Mechanical and software overload protection minimizes damage to weighing cell.
$234.27 - $277.00CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (g) Power (VAC) Readability (g)
Availability Pricing
Ohaus CS200-001 Portable Compact Scale, 200g x 0.1g RK-11003-02 Mfr # CS200
COMPARE
200 115 0.1
$234.27CAD / Each
Ohaus CS-2000 Portable Compact Scale, 2000g x 1g RK-11003-12 Mfr # CS2000
COMPARE
2000 115 1
$234.27CAD / Each
Ohaus CS-5000 Portable Compact Scale, 5000g x 1g RK-11003-22 Mfr # CS5200
COMPARE
5000 115 1
$277.00CAD / Each