Oakton® Waterproof DO 450 Optical Dissolved Oxygen Portable Meter
Affordable optical RDO technology reduces maintenance time and costs
  • Eliminates costly, tiresome membranes and solutions
  • Rugged, IP67-rated housing for plant and field applications
  • Memory for 500 data sets and USB and RS-232 output for tranfer to PC
  • 500 hours battery life, or optional universal power adapter
  • Built-in stand with wall mount allows for easy benchtop measurement or storage
The DO 450 meter uses optical measurement technology offeringfast response times and long-term stability with minimal maintenance. Plus automatic temperature compensation (ATC), barometric pressure compensation, and salinity correction to further ensure measurement accuracy. The large, backlit LCD lets you view % saturation or mg/L (ppm) simultaneously with temperature.
Rugged, IP67-rated waterproof housing makes the meter ideal for plant or field testing. Additional features include ready (stability) indicator, smart averaging, hold function, calibration alarm, and battery-life indicator.
Perform data logging with manual or timed collection of up to 500 data sets with real-time clock time/date stamp. Order optional data cables for USB and RS-232 connectivity.
5 PRODUCT OPTIONS
Compareitem specifications
Min Dissolved Oxygen Description
Availability Pricing
COMPARE
0 to 20 ppm; 0 to 200% Waterproof Portable Meter with Probe
$2,725.77CAD / Each
COMPARE
0 to 20 ppm; 0 to 200% Waterproof Meter with Probe with Calibration
$2,360.37CAD / Each
COMPARE
0 to 20 ppm; 0 to 200% Waterproof Portable Meter Only
$1,183.13CAD / Each
COMPARE
0 to 20 ppm; 0 to 200% Waterproof Portable Meter Kit
$3,315.13CAD / Each
COMPARE
0 to 20 ppm; 0 to 200% Waterproof Portable Meter Kit with Calibration
$2,391.31CAD / Each